Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MED28, Mouse, Unlabeled, Clone: 7E1,

Catalog No. MABC1143MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC1143MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1143MI Supplier MilliporeSigma™ Supplier No. MABC1143
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mediator of RNA polymerase II transcription subunit 28 (UniProt Q9H204; also known as Endothelial-derived protein 1, Magicin, Mediator complex subunit 28, Merlin and Grb2-interacting cytoskeletal protein, Tumor angiogenesis marker EG-1) is encoded by the MED28 (also known as EG1, FKSG20) gene (Gene ID 80306) in human. MED28 (Magicin) is a subunit of the Mediator complex that is composed of at least 30 subunits and plays an essential role in transcriptional regulation by bridging RNA polymerase II transcriptional machinery and DNA-bound regulatory factors. MED28 is reported to mediate cell surface receptor-mediated signaling and associate with Src-family kinases (e.g. Fyn, Src and Lck), as well as with the Neurofibromatosis 2 (NF2) tumor suppressor merlin. MED28 is associated with both the cytoskeleton and the nucleus, indicating a role as an adaptor/scaffolding protein to relay cellular signals to the cytoskeleton and the nucleus. Homozygous Med28-deletion causes peri-implantation lethality with a loss of pluripotency within the inner cell mass (ICM) of the blastocyst. In addition, Med28-/- embryonic fibroblasts (MEFs) lose the capability to reprogram and Med28 overexpression augments reprogramming efficiency. Upregulated MED28 expression is associated with poor outcome in breast cancer patients, and targeted MED28 downregulation decreases breast cancer cell proliferation and tumor growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED28
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7E1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene MED28, EG1, FKSG20
Gene Accession No. Q9H204
Host Species Mouse
Immunogen GST-tagged recombinant human MED28 lacking N-terminal 12 amino acids.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_079481
Test Specificity Clone 7E1 detected MED28 in wild-type, but not Med28-/- murine cells. Clone 7E1 detected Cre expression-induced MED28 knockdown in MEFs with floxed Med28 alleles (Li, L., et al. (2015). PLoS One. 10(10):e0140192).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.