Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Antiseptic Pump

Catalog No. 19090658 Shop All Medique Products
Change view
Click to view available options
Volume (English):
2 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-090-658 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-658 Supplier Medique Supplier No. 25702
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For minor cuts and abrasions.

  • Contains Hydrogen peroxide

Specifications

Product Type Antiseptic Pump
For Use With (Application) For minor cuts and abrasions
Volume (English) 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.