Learn More
Medique Medi-First™ Antiseptic Wipes
Catalog No. 19090836 Shop All Medique Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-836 100/Pk.
19-090-835 20/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-090-836 Supplier Medique Supplier No. 21433
Add to Cart
Add to Cart
Disinfects and cleans skin as first-aid antiseptic.
  • Towelettes saturated with 1:750 Benzalkonium chloride solution
Specifications
For Use With (Application) Disinfects, cleans skin as first-aid antiseptic
Product Type Antiseptic Wipe
Description Individual tamper-evident packets
Material 1:750 Benzalkonium chloride solution
Packaging Individual towelettes
Quantity 100/Pk.