Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Blood Clotting Spray

Catalog No. 19090653 Shop All Medique Products
Change view
Click to view available options
Includes:
3 oz can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-090-653 3 oz can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-653 Supplier Medique Supplier No. 22617

missing translation for 'validMainframeMsg'

Stops superficial bleeding on contact.

  • Assists in fighting infection
  • Temporarily relieves pain from minor cuts, burns and scrapes

Specifications

Product Type Blood Clotting Spray
For Use With (Application) Stops superficial bleeding on contact
Format Aerosol
Includes 3 oz can
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.