Learn More
Medique Medi-First™ Chemical Burn Kit
Catalog No. 19090629 Shop All Medique Products
This item is not returnable. View return policy
Catalog No. 19-090-629 Supplier Medique Supplier No. 89612
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Two ABD Sterile Pads 5 x 9 in., Two ABD Sterile Pads 8 x 10 in., one Adhesive Tape, 1/2 in. x 5 yards, Two Vinyl Gloves, pair, one EMT Utility Scissors 7 in., Two Calgonate for Acid Burns 25g, Two 2 in. Stretch Roller Gauze, two 3 in. Stretch Roller Gauze
Consists of products required to provide quick treatment for chemical burns.
  • Kit contains ABD sterile pads, Adhesive Tape, Vinyl Gloves, EMT Utility Scissors, Calgonate for Acid Burns, Stretch Roller Gauze, large Cold Pack, Antiseptic Spray, Medi-Wash Eyewash, Eye Pads, Penlight, Ammonia Wipes, Gauze Pads
Specifications
Product Type Chemical Burn Kit
Includes Two ABD Sterile Pads 5 x 9 in., Two ABD Sterile Pads 8 x 10 in., one Adhesive Tape, 1/2 in. x 5 yards, Two Vinyl Gloves, pair, one EMT Utility Scissors 7 in., Two Calgonate for Acid Burns 25g, Two 2 in. Stretch Roller Gauze, two 3 in. Stretch Roller Gauze
Dimensions (L x W x H) 9.12 x 9.12 x 2.75 in.