Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Ibuprofen Tablets

Catalog No. 19090795 Shop All Medique Products
Change view
Click to view available options
Quantity:
2/Pk., 125/Box
2/Pk., 250/Box
2/Pk., 50/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-795 2/Pk., 50/Box
19-090-794 2/Pk., 125/Box
19-090-793 2/Pk., 250/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-795 Supplier Medique Supplier No. 80833
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For fast and effective relief of headache, muscular aches, arthritis pain and menstrual cramps.

  • Helps reduce fever
  • Each tablet contains Ibuprofen 200mg

Specifications

For Use With (Application) Fast and effective relief of headache, muscular aches, arthritis pain and menstrual cramps
Active Ingredients Ibuprofen (200 mg)
Product Type Ibuprofen
Quantity 2/Pk., 50/Box
Recommended Age 12 years and older
Storage Requirements 68°F to 77°F (20°C to 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.