Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Instant Cold Packs

Catalog No. 19090709 Shop All Medique Products
Change view
Click to view available options
Size:
Small
Large
Dimensions (L x W):
4 x 6 in.
5 x 9 in.
Format:
Boxed
Unboxed
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Dimensions (L x W) Format
19-090-709 Large 5 x 9 in. Unboxed
19-090-708 Large 5 x 9 in. Boxed
19-090-710 Small 4 x 6 in. Boxed
19-090-711 Small 4 x 6 in. Unboxed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-090-709 Supplier Medique Supplier No. 7021M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For fast pain relief.

  • Instant cold compress

Specifications

For Use With (Application) For fast pain relief
Product Type Instant Cold Pack
Format Unboxed
Size Large
Dimensions (L x W) 5 x 9 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.