Learn More
Medique Medi-First™ Instant Hot Packs
Catalog No. 19090707 Shop All Medique Products
Change view
Click to view available options
Format:
Boxed
Unboxed
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19-090-707 Unboxed
19-090-706 Boxed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-090-707 Supplier Medique Supplier No. 7131M
Add to Cart
Add to Cart
Single use only. Medi-Fast Instant Hot Packs are convenient for first aid kits.
  • Activated simply by squeezing to break the inner pouch and shaking it until it starts to get warm
  • Disposable
Specifications
Product Type Instant Hot Pack
Disposable Single-use
Format Unboxed
Size Large
Dimensions (L x W) 5 x 9 in.