Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Metal Detectable Blue Bandages

Catalog No. 19090586 Shop All Medique Products
Change view
Click to view available options
Style:
Plastic Strip
Woven Strip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
19-090-586 Plastic Strip
19-090-585 Woven Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-090-586 Supplier Medique Supplier No. 67133
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifically developed for use in food and restaurant preparations.

  • Designed to be highly visible
  • Latex-free

Specifications

For Use With (Application) food and restaurant preparations
Product Type Metal-detectable Bandage
Color Blue
Latex Free Yes
Quantity 100 Pk.
Style Plastic Strip
Dimensions (L x W) 1 x 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.