Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Non-Aspirin Tablets

Catalog No. 19090798 Shop All Medique Products
Change view
Click to view available options
Quantity:
2/Pk., 125/Box
2/Pk., 250/Box
2/Pk., 50/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-798 2/Pk., 50/Box
19-090-797 2/Pk., 125/Box
19-090-796 2/Pk., 250/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-798 Supplier Medique Supplier No. 80333
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the relief of fever, headache, minor muscular aches, arthritis pain.

  • Gentler on stomach than aspirin
  • Each tablet contains Acetaminophen 325mg

Specifications

Product Type Non-aspirin Pain Reliever
For Use With (Application) For pain, ache, and fever relief
Active Ingredients Acetaminophen (325 mg)
Quantity 2/Pk., 50/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.