Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Pain and Pressure Tablets

Catalog No. 19090818 Shop All Medique Products
Change view
Click to view available options
Quantity:
2/Pk., 125/Box
2/Pk., 250/Box
2/Pk., 50/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-818 2/Pk., 50/Box
19-090-817 2/Pk., 125/Box
19-090-816 2/Pk., 250/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-818 Supplier Medique Supplier No. 81933
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Relieves sinus pressure associated with allergies, headaches and colds.

  • Easy to swallow and film-coated
  • Each tablet contains: Acetaminophen 500mg; Phenylephrine HCl 5mg

Specifications

Active Ingredients Acetaminophen (500 mg), Phenylephrine HCl (5 mg)
For Use With (Application) Relieves sinus pressure associated with allergies, headaches and colds
Product Type Pain and Pressure Tablets
Format Film-coated Tablet
Quantity 2/Pk., 50/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.