Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Sterile Gauze Pads

Catalog No. 19090736 Shop All Medique Products
Change view
Click to view available options
Quantity:
10 Pk.
100 Pk.
25/Box
Dimensions (L x W):
2 x 2 in.
3 x 3 in.
4 x 4 in.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dimensions (L x W)
19-090-736 25/Box 4 x 4 in.
19-090-727 10 Pk. 2 x 2 in.
19-090-728 25/Box 2 x 2 in.
19-090-729 100 Pk. 2 x 2 in.
19-090-732 10 Pk. 3 x 3 in.
19-090-734 100 Pk. 3 x 3 in.
19-090-733 25/Box 3 x 3 in.
19-090-735 10 Pk. 4 x 4 in.
19-090-737 100 Pk. 4 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-090-736 Supplier Medique Supplier No. 62073
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sterile gauze pads use to clean and cover wounds, also for use as a wet absorbent dressing. Individually wrapped super absorbent sterile pads.

  • Latex-free
  • To clean and cover wounds
  • Also used as wet absorbent dressing

Specifications

For Use With (Application) For minor first aid and medical needs
Product Type Gauze Pad
Sterility Sterile
Latex Free Yes
Quantity 25/Box
Dimensions (L x W) 4 x 4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.