Learn More
Medique Medi-First™ Triple Antibiotic Ointment
Catalog No. 19090844 Shop All Medique Products
Change view
Click to view available options
Quantity:
10/Box
144/Box
25/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-844 25/Box
19-090-832 10/Box
19-090-845 144/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-844 Supplier Medique Supplier No. 22373
Only null left
Add to Cart
Add to Cart
For effective protection against infection from minor cuts, burns and abrasions.
  • Active ingredients: Bacitracin Zinc (400 units Bacitracin); Neomycin Sulfate 5mg; Polymyxin-B Sulfate 5000 units
Specifications
For Use With (Application) Protection against infection from minor cuts, burns, abrasions
Active Ingredients Bacitracin Zinc (400U), Neomycin Sulfate (5 mg), Polymyxin B Sulfate (5000U)
Product Type Triple Antibiotic Ointment
Quantity 25/Box