Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Rubber Cap/Liner
Shop All DWK Life Sciences Products
Description
- Manufactured from borosilicate glass which provides the best chemical resistance
- Clear borosilicate glass bottle
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Black Phenolic Rubber Lined Cap
- Hazardous communication label included with each case of bottles allows for content ID and safety notification
- Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Clear |
| Material | Borosilicate Glass (Type I) |
| Quantity | 48 EA |
| Closure Color | Black |
| Product Type | Media Bottle |
| Closure Liner | Rubber |
| Closure Material | Phenolic |