Learn More
Mediator complex subunit 25 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Mediator complex subunit 25 |
| Applications | Western Blot, Inhibition Assays, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | MED25,ACID1,ARC92,PTOV2,TCBAP0758 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 16 amino acids from the internal region of human Mediator of RNA polymerase II transcription subunit 25 (MED25). |
| Purification Method | Affinity Purified |
| Quantity | 25 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.