Learn More
Medica Urine Diluent For EasyLyte Analyzers
SDP
Catalog No. 22515378 Shop All Medica Corporation Products
Click to view available options
For Use With (Equipment):
EasyLyte Analyzers
Catalog No. For Use With (Equipment)
22-515-378 EasyLyte Analyzers
1 options
Catalog No. 22-515-378 Supplier Medica Supplier No. 2111
ISE diluent for urinalysis
  • Volume: 500mL
Specifications
Type Urine Diluent
For Use With (Equipment) EasyLyte Analyzers
Capacity (Metric) 500 mL