Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Biohazardous Waste Bags, Autoclavable
SDP

Catalog No. 22023750 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
35 x 25 in. (88.9 x 63.5cm)
38 x 47 in.
40 x 70 in. (101.6 x 177.8cm)
Color:
Clear
Orange, Black
Red, Black
Thickness (Metric):
0.045 mm
0.051 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color Thickness (Metric)
22-023-750 40 x 70 in. (101.6 x 177.8cm) Red, Black 0.051 mm
14-828-350 38 x 47 in. Clear 0.045 mm
22-023803 35 x 25 in. (88.9 x 63.5cm) Orange, Black 0.045 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-023-750 Supplier Medegen Supplier No. 5028
Add to Cart
Add to Cart
This item is not returnable. View return policy

These Biohazardous Waste Bags are autoclavable.

  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Meets 165 g Dart Impact Resistance Test
Color Red, Black
Includes Indicator, flat pack, twist ties
Material Polypropylene
Thickness (English) 2 mil
Dimensions (L x W) 40 x 70 in. (101.6 x 177.8cm)
Length (English) 40 in.
Width (English) 70 in.
Product Type Autoclavable Biohazardous Waste Bag
Capacity (English) 55 to 60 gal.
Capacity (Metric) 208.2 to 227.1 L
Length (Metric) 101.6 cm
Width (Metric) 177.8 cm
Thickness (Metric) 0.051 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.