Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Biohazardous Waste Bags, HDPE Film, Coreless Roll
SDP

Catalog No. 14828230 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
30.5 x 43 in.
24 x 24 in.
24 x 33 in.
38 x 46 in.
40 x 48 in.
Capacity (English):
12 to 16 gal.
30 to 32 gal.
40 to 45 gal.
44 gal.
8 to 10 gal.
Thickness (Metric):
2.5 μm
2.8 μm
3.6 μm
4.1 μm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Capacity (English) Thickness (Metric)
14-828-230 24 x 33 in. 12 to 16 gal. 2.8 μm
14-828-227 24 x 24 in. 8 to 10 gal. 2.8 μm
14-828-236 30.5 x 43 in. 30 to 32 gal. 3.6 μm
14-828-250 38 x 46 in. 44 gal. 4.1 μm
14-828-260 40 x 48 in. 40 to 45 gal. 2.5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-828-230 Supplier Medegen Supplier No. RS243311RH
Add to Cart
Edge
Add to Cart

Meet ASTM™ Dart test requirements.

  • Red bag with black printing
  • Biohazardous/Infectious Waste label with Biohazard symbol printed on bag

Specifications

Certifications/Compliance ASTM Dart
Color Red, Black
Includes Biohazardous or infectious waste with biohazard symbol
Material HDPE
Thickness (English) 0.11 mil
Dimensions (L x W) 24 x 33 in.
Length (English) 24 in.
Width (English) 33 in.
Product Type Biohazardous Waste Bag
Capacity (English) 12 to 16 gal.
Capacity (Metric) 45.4 to 60.6 L
Length (Metric) 60.96 cm
Width (Metric) 83.82 cm
Thickness (Metric) 2.8 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.