Learn More
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
Shop All American Precision Plastics Products

Description
- Red bag with black printing
- Biohazardous/Infectious Waste label with Biohazard symbol printed on bag
Specifications
Specifications
| Certifications/Compliance | ASTM Dart |
| Color | Red, Black |
| Includes | Biohazardous or infectious waste with biohazard symbol |
| Material | LLDPE |
| Thickness (English) | 1.75 mil |
| Dimensions (L x W) | 38 x 45 in. |
| Length (English) | 38 in. |
| Width (English) | 45 in. |
| Product Type | Biohazardous Waste Bag |
| Capacity (English) | 44 gal. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.