Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
GSA_VASDP

Catalog No. 14828223 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
14.5 x 19 in.
11 x 14.25 in.
16 x 24 in.
23 x 23 in.
25 x 34 in.
38 x 45 in.
Capacity (English):
1 to 3 gal.
12 to 16 gal.
2 to 3 gal.
44 gal.
5 to 6 gal.
7 to 10 gal.
Thickness (English):
1.20 mil
1.25 mil
1.50 mil
1.75 mil
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Capacity (English) Thickness (English)
14-828-223 14.5 x 19 in. 2 to 3 gal. 1.25 mil
14-828-197 23 x 23 in. 7 to 10 gal. 1.50 mil
14-828-222 11 x 14.25 in. 1 to 3 gal. 1.25 mil
14-828-224 16 x 24 in. 5 to 6 gal. 1.25 mil
14-828-192 25 x 34 in. 12 to 16 gal. 1.20 mil
14-828-194 38 x 45 in. 44 gal. 1.75 mil
14-828-201 38 x 45 in. 44 gal. 1.20 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-828-223 Supplier Medegen Supplier No. RD630
Add to Cart
Edge
Add to Cart

Meet ASTM™ Dart/Elmendorf and DOT CFR 49 173-197 requirements.

  • Red bag with black printing
  • Biohazardous/Infectious Waste label with Biohazard symbol printed on bag

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance ASTM Dart
Color Red, Black
Includes Biohazardous or infectious waste with biohazard symbol
Material LLDPE
Thickness (English) 1.25 mil
Dimensions (L x W) 14.5 x 19 in.
Length (English) 14.5 in.
Width (English) 19 in.
Product Type Biohazardous Waste Bag
Capacity (English) 2 to 3 gal.
Capacity (Metric) 7.6 to 11.4 L
Length (Metric) 36.83 cm
Width (Metric) 48.26 cm
Thickness (Metric) 0.032 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.