Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Facility Safety and Maintenance
- Waste Disposal
- Hazardous and Biohazardous Waste Disposal Bags and Containers
- Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
Catalog No. 14828109
Shop All American Precision Plastics Products

Change view
Click to view available options
Dimensions (L x W):
30.5 x 41 in.
23 x 23 in.
24 x 30 in.
25 x 34 in.
33 x 40 in.
37 x 50 in.
38 x 45 in.
40 x 45 in.
40 x 46 in.
43 x 48 in.
43 x 55 in.
Capacity (English):
12 to 16 gal.
20 to 30 gal.
30 gal.
33 gal.
40 to 45 gal.
44 gal.
55 gal.
7 to 10 gal.
8 to 12 gal.
Thickness (English):
1.1 mil
1.2 mil
1.3 mil
1.4 mil
1.5 mil
1.75 mil
2 mil
3 mil
Certifications/Compliance:
ASTM Dart
ASTM Dart or Elmendorf, DOT CFR 49 173-197
24 product options available for selection
| Catalog No. | Dimensions (L x W) | Capacity (English) | Thickness (English) | Certifications/Compliance |
|---|---|---|---|---|
| 14-828-109 | 43 x 55 in. | 55 gal. | 3 mil | ASTM Dart |
| 14-828-335 | 25 x 34 in. | 12 to 16 gal. | 1.2 mil | ASTM Dart |
| 14-828-326 | 23 x 23 in. | 7 to 10 gal. | 1.1 mil | ASTM Dart |
| 14-828-85 | 33 x 40 in. | 33 gal. | 1.5 mil | ASTM Dart |
| 14-828-90 | 38 x 45 in. | 44 gal. | 1.75 mil | ASTM Dart |
| 14-828-89 | 38 x 45 in. | 44 gal. | 1.5 mil | ASTM Dart |
| 14-828-327 | 23 x 23 in. | 7 to 10 gal. | 1.5 mil | ASTM Dart |
| 14-828-96 | 30.5 x 41 in. | 30 gal. | 2 mil | ASTM Dart |
| 14-828-328 | 30.5 x 41 in. | 30 gal. | 1.5 mil | ASTM Dart |
| 14-828-329 | 30.5 x 41 in. | 30 gal. | 3 mil | ASTM Dart |
| 14-828-98 | 38 x 45 in. | 44 gal. | 3 mil | ASTM Dart |
| 18-023-851 | 38 x 45 in. | 44 gal. | 1.2 mil | ASTM Dart |
| 14-828-331 | 43 x 48 in. | 55 gal. | 2 mil | ASTM Dart |
| 14-828-108 | 43 x 55 in. | 55 gal. | 1.5 mil | ASTM Dart |
| 14-828-158 | 23 x 23 in. | 7 to 10 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-157 | 37 x 50 in. | 44 gal. | 1.5 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-156 | 38 x 45 in. | 44 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-161 | 24 x 30 in. | 8 to 12 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-159 | 30.5 x 41 in. | 20 to 30 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-163 | 37 x 50 in. | 44 gal. | 1.3 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-164 | 40 x 46 in. | 40 to 45 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-827-43B | 30.5 x 41 in. | 30 gal. | 1.2 mil | ASTM Dart |
| 14-828-166 | 40 x 45 in. | 40 to 45 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-336 | 23 x 23 in. | 7 to 10 gal. | 1.4 mil | ASTM Dart |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
Description
- Made from low-density polyethylene
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
- Individually folded
Specifications
Specifications
| Certifications/Compliance | ASTM Dart |
| Color | Red, Black |
| For Use With (Application) | Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab |
| Includes | Biohazardous or infectious waste with biohazard symbol |
| Material | LLDPE |
| Thickness (English) | 3 mil |
| Dimensions (L x W) | 43 x 55 in. |
| Length (English) | 43 in. |
| Width (English) | 55 in. |
| Product Type | Biohazardous Waste Bag |
| Capacity (English) | 55 gal. |
| Capacity (Metric) | 208.2 L |
| Length (Metric) | 109.22 cm |
| Width (Metric) | 139.7 cm |
| Thickness (Metric) | 0.076 mm |
| Show More Show Less |
May NOT be used in CA, FL, PA or for interstate (between state) transport.
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?