Learn More
Description
- Rectangular, easy-to-assemble container stacks for convenient storage until incinerated
- Integral handles for safe handling
- Unassembled containers store flat to conserve space
Specifications
Specifications
| Capacity (Metric) | 9.5 L |
| Design | Infectious Waste with Biohazard Symbol |
| Diameter (Metric) Outer | 15.24 cm |
| Diameter (English) Outer | 6 in. |
| Capacity (English) | 2.5 gal. |
| Color | Red, Black |
| Height (Metric) | 26.6 cm |
| Width (Metric) | 30.4 cm |
| Liner Material | Plastic |
| Width (English) | 12 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
