Learn More
Cliawaived Inc N3100 Series Nitrile Exam Gloves Medium. Powder free, exam grade, Chemo tested (1 box of 100 gloves)
SDP
Supplier:  Cliawaived Inc VAL3100M
Encompass_Preferred
N3100 Series Nitrile Exam Gloves Medium. Powder free, exam grade, Chemo tested. Dependable for house-wide applications within Healthcare, Labs, EMS and other settings where high-use and disposable exam glove protection is important. Size: Extra LargeNitrileThickness (Finger): 3.5milPowder FreeASTM D6319, D6978Exam GradeColor: BlueTexture: FingertipLength: Standard (240mm)Cuff: BeadedNon-SterileChemotherapy Tested**Chemotherapy drug testing information on back of box(1 box of 100)
Catalog No. NC2580180
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.