Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medikoff™ Cough Drops

Catalog No. 19090768 Shop All Medique Products
Change view
Click to view available options
Quantity:
600 Drops
600 Drops/Jar
75 Drops
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-768 600 Drops
19-090-767 75 Drops
19-090-766 600 Drops/Jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-768 Supplier Medique Supplier No. 50601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For relief from throat irritation and coughs and to clear nasal passages.

  • Each tablet contains: Menthol 7.6mg

Specifications

Active Ingredients Menthol (7.6 mg)
For Use With (Application) For relief from throat irritation and coughs and to clear nasal passages
Product Type Cough Drops
Color Red
Flavor Cherry
Quantity 600 Drops
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.