Learn More
INTRAVENOUS QUALITY ASSURANCE (IVQA) Category 2 CSP Media-Fill Test Kit
Catalog No. NC0561157
Encompass_Preferred
The Category 2 CSP Media-Fill Test Kit validates compounding personnel's aseptic technique and sterile compounding proficiency. Intended for simulating Category 2 compounded sterile preparations, it evaluates the risk of microbial contamination and assesses competency in aseptic techniques. Suitable for the initial assessment and every 6-month revalidation of aseptic manipulation competency, in accordance with USP Chapter <797> guidelines.Kit Contains:
•3 - Sterile 50ml (TSB) Test Media Vials•6 - Sterile 50ml Empty Serum Vials•3 - Sterile 10ml Empty Serum Vials•1 - Whirl-Pak Bag•1 - Log Sheet
Catalog No. NC0561157 Supplier INTRAVENOUS QUALITY ASSURANCE (IVQA) Supplier No. MFT02
The Category 2 CSP Media-Fill Test Kit validates compounding personnel's aseptic technique and sterile compounding proficiency. Intended for simulating Category 2 compounded sterile preparations, it evaluates the risk of microbial contamination and assesses competency in aseptic techniques. Suitable for the initial assessment and every 6-month revalidation of aseptic manipulation competency, in accordance with USP Chapter <797> guidelines.Kit Contains:
•3 - Sterile 50ml (TSB) Test Media Vials•6 - Sterile 50ml Empty Serum Vials•3 - Sterile 10ml Empty Serum Vials•1 - Whirl-Pak Bag•1 - Log Sheet