Learn More
Description
- Polyester tags provide identification solutions for almost any environment
- Tag includes a write-on area that accepts most marking pens
- Tags are appropriate for indoor or outdoor use
- Tags are water-resistant
Specifications
Specifications
| Quantity | 25/Pk. |
| Color | Yellow |
| Material | Polyester |
| Includes | (25) nylon fasteners |
| Shape | Rectangle |
| Waterproof | Water-resistant |
| Dimensions (L x W) | 2 x 4 in. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.