Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Medroxyprogesterone 17-acetate, Progesterone Receptor Agonist, Solid, 500 mg

Catalog No. 502706212
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6212 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6212 Supplier Abcam Supplier No. AB142633500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic variant of progesterone and potent progesterone receptor agonist with angiostatic properties.

  • Molecular Weight: 386.5 Da
  • Purity: >99%
  • Form: Solid
  • Activity: Potent progesterone receptor agonist; active in vitro and in vivo
  • Biological Function: Angiostatic steroid; inhibits vascularization, collagenolysis, tumor growth, and plasmin generation
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 99.74% Medroxyprogesterone Acetate
Content And Storage Store at -20°C.
Shipping Condition Ambient
Target Molecule Progesterone Receptor
Form Solid
Formulation 99.74% Medroxyprogesterone Acetate
Quantity 500 mg
For Use With (Application) Angiostatic research, tumor growth inhibition, plasmin generation inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.