Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Medroxyprogesterone 17-acetate, Progesterone Receptor Agonist, Solid, >99% Purity, 100 mg
Description
- Purity: >99%
- Molecular Weight: 386.5 Da
- Form: Solid
- Properties: Synthetic variant of progesterone and potent progesterone receptor agonist; angiostatic steroid that inhibits vascularization, collagenolysis, tumor growth, and plasmin generation
- Activity: Active in vitro and in vivo
- CAS Number: 71-58-9
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Progesterone Receptor |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Quantity | 100 mg |
| For Use With (Application) | Progesterone receptor signaling study, Angiogenesis inhibition, Tumor growth inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?