Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEF2C Monoclonal Antibody (OTI4F7)
GREENER_CHOICE

Catalog No. PIMA525477 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525477 Supplier Invitrogen™ Supplier No. MA525477
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEF2C Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

MEF2C is a transcription activator which binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. Controls cardiac morphogenesis and myogenesis, and is also involved in vascular development. Plays an essential role in hippocampal-dependent learning and memory by suppressing the number of excitatory synapses and thus regulating basal and evoked synaptic transmission. Crucial for normal neuronal development, distribution, and electrical activity in the neocortex. Necessary for proper development of megakaryocytes and platelets and for bone marrow B lymphopoiesis. Required for B-cell survival and proliferation in response to BCR stimulation, efficient IgG1 antibody responses to T-cell-dependent antigens and for normal induction of germinal center B cells. May also be involved in neurogenesis and in the development of cortical architecture. Isoform 3 and isoform 4, which lack the repressor domain, are more active than isoform 1 and isoform 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2C
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MEF2C
Gene Accession No. Q06413
Gene Alias 5430401D19Rik; 9930028G15Rik; AV011172; C5DELq14.3; DEL5q14.3; hoo; hoover; id:ibd5026; MADS box transcription enhancer factor 2, polypeptide C; Mef2; mef2c; mef2c protein; mef2ca; Myocyte enhancer factor 2C; myocyte enhancer factor 2C MEF2C; myocyte enhancer factor 2ca; myocyte enhancer factor 2ca 4'-6'; myocyte enhancer factor 2ca delta gamma-like; myocyte enhancer factor 2ca variant 4; myocyte enhancer factor 2ca variant 5; myocyte enhancer factor 2ca variant 6; myocyte enhancer factor 2ca; myocyte-specific enhancer factor 2C; myocyte-specific enhancer factor 2C; RGD1563119; wu:fc05b06; zgc:64184; zgc:85726
Gene Symbols MEF2C
Host Species Mouse
Immunogen Full length human recombinant protein of MEF2C produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4208
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.