Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mefenamic Acid 98.0+%, TCI America™
SDP

Catalog No. M1782100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1782100G 100 g
M178225G 25 g
M1782500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1782100G Supplier TCI America Supplier No. M1782100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61-68-7
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD00051721
InChI Key HYYBABOKPJLUIN-UHFFFAOYSA-N
Synonym mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid
PubChem CID 4044
ChEBI CHEBI:6717
IUPAC Name 2-(2,3-dimethylanilino)benzoic acid
SMILES CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 100 g
Formula Weight 241.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mefenamic Acid
EINECSNumber (3)-1469
RTECSNumber CB4550000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.