Learn More
PIP™ Mega Bullet™ BioSoft™ Ear Plug
GREENER_CHOICE
Catalog No. 17467655 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Corded:
Corded
Uncorded
Packaging Type:
Bag
Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Packaging Type
17-467-655 Uncorded Bag
17-467-656 Uncorded Box
17-467-654 Corded Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-467-655 Supplier PIP™ Supplier No. BSF1B
Only null left
Add to Cart
Add to Cart
Disposable, bio-based foam ear plug with BioSoft™ Technology.
  • PIP™ understands the importance of reducing waste to lower costs and minimize environmental impact
  • New, sustainable ECOSeries™ BioSoft™ ear plugs are made from bio-based technologies
  • BioSoft™ ear plug foam material provides the same fit and performance as conventional polyurethane or PVC materials, but with a lower carbon footprint
  • Innovative technology not only reduces emissions during manufacturing, but contains bio-based materials, making BioSoft™ more environmentally friendly at end of use
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
High Visibility No
Packaging Type Bag
Quantity 2000 Plugs/Cs.
Noise Reduction Rating (NRR) 32 NRR
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Dielectric Yes
EN 352.2002 EN 352-1
Includes Ear Plugs