Learn More
PIP™ Mega Bullet™ Plus Disposable Soft Polyurethane Foam Ear Plugs - NRR 33
Catalog No. 17489664 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
33
Catalog No. Noise Reduction Rating (NRR)
17-489-664 33
1 options
Catalog No. 17-489-664 Supplier PIP™ Supplier No. 267HPF810
  • $48.75
    $27.65 / Pack of 200
    Save $21.10
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Disposable soft polyurethane foam ear plugs - NRR 33.
  • PowerSoft™ Foam Technology features a proprietary polyurethane formulation that delivers optimal recovery time, easy insertion and maximum all-day comfort
  • Size and shape fits most ear canals
  • Smooth closed cell outer layer that prevents debris build-up
  • Soft and comfortable PowerSoft™ foam supports long term comfort
  • Black PVC cord for convenience and to prevent loss of ear plugs
  • NRR 33 Protection
Specifications
Size Universal
Noise Reduction Rating (NRR) 33
Corded Uncorded
Plug Shape Bullet
Product Type Disposable Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric Yes