Learn More
PIP™ Mega Bullet™ Plus Prefilled Mega Bullet™ Plus Ear Plug Dispenser - 500 Pairs
Catalog No. 17489667 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
33
Catalog No. Noise Reduction Rating (NRR)
17-489-667 33
1 options
Catalog No. 17-489-667 Supplier PIP™ Supplier No. 267HPD810500
  • $147.00
    $84.65 / Each of 1
    Save $62.35
    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Prefilled mega bullet™ plus ear plug dispenser - 500 pair.
  • Prefilled dispenser includes 500 pairs of Mega Bullet™ Plus ear plugs
  • Table top install and easy to use
  • Controlled release mechanism ensures the correct number of plugs are dispensed into a collection tray
  • Sufficient capacity to supply large production areas
  • Transparent design to monitor the number of earplugs remaining
Specifications
Size Universal
Noise Reduction Rating (NRR) 33
Corded Corded
Plug Shape Bullet
Product Type Ear Plug Dispenser with PD500 Plugs
Disposable Single-use
Metal Detectable No
Dielectric No
Material Plastic
Style Prefilled
Mounting Flat Surface, Wall Mountable
Foldable No
Includes Prefilled Display Dispenser