Learn More
PIP™ Mega Flare™ Plus Disposable Soft Polyurethane Foam Ear Plugs - NRR 33
Catalog No. 17489662 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
33
Catalog No. Noise Reduction Rating (NRR)
17-489-662 33
1 options
Catalog No. 17-489-662 Supplier PIP™ Supplier No. 267HPF910
Add to Cart
Add to Cart
Disposable soft polyurethane foam ear plugs - NRR 33.
  • Consistent cellular structure for optimal expansion recovery time for a custom, uniform fit
  • Smooth closed cell outer layer that prevents debris build-up
  • Soft and comfortable PowerSoft™ foam supports all day comfort
  • NRR 33 protection
Specifications
Size Universal
Noise Reduction Rating (NRR) 33
Corded Uncorded
Plug Shape Bell
Product Type Disposable Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric Yes