Learn More
PIP™ Mega Flare™ Soft Polyurethane Foam Ear Plugs
Catalog No. 19046462 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
32 dB
Catalog No. Noise Reduction Rating (NRR)
19-046-462 32 dB
1 options
Catalog No. 19-046-462 Supplier PIP™ Supplier No. 267HPF410C
Add to Cart
Add to Cart
Mega Flare™ disposable ear plugs have a traditional flared bell shape for easy removal.
  • Consistent cellular structure for optimal expansion recovery time for a consistent, uniform fit
  • Smooth closed cell outer layer prevents debris build-up
  • Soft and comfortable feel for all day comfort
  • Black PVC cord for convenience and to prevent loss of ear plugs
  • NRR 32 protection
Specifications
ANSI S3.1974 Yes
Cord Color Black
Cord Material PVC
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Oil and gas, Manufacturing, Workshops, Building, Construction, Energy
High Visibility Yes
Packaging Type Poly bag
Plug Material Polyurethane foam
Plug Color Red
Quantity 100/Pack; 10 Packs/Case
Noise Reduction Rating (NRR) 32 dB
Corded Corded
Plug Shape Bell
Product Type Corded Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric Yes
EN 352.2002 EN 352-2