Learn More
PIP™ Mega T-Fit™ Soft Polyurethane Foam Ear Plugs
Catalog No. 19046463 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
32 dB
Catalog No. Noise Reduction Rating (NRR)
19-046-463 32 dB
1 options
Catalog No. 19-046-463 Supplier PIP™ Supplier No. 267HPF510
Add to Cart
Add to Cart
Mega T-Fit™ disposable ear plugs are T-shaped for easy removal.
  • Consistent cellular structure for optimal expansion recovery time for a consistent, uniform fit
  • Smooth closed cell outer layer prevents debris build-up
  • Soft and comfortable PowerSoft™ foam supports all day comfort
  • NRR 32 protection
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Oil and gas, Manufacturing, Workshops, Building, Construction, Energy
High Visibility Yes
Packaging Type Poly bag
Pairs per Package 200 pairs/dispenser box
Plug Material Polyurethane foam
Plug Color Orange
Noise Reduction Rating (NRR) 32 dB
Corded Uncorded
Plug Shape T-Shape
Product Type Disposable Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric Yes
EN 352.2002 EN 352-2