Learn More
Leica Biosystems MEGACASSETTES GREEN (1000)
Supplier:  Leica Biosystems 38VSP59045
Encompass
These cassettes offer double the depth of conventional cassettes.Allows for easier embedding and processing of larger or whole organs (such as eyes, teeth and bones), as well as ossified tissue that do not fit standard cassettes.Perfect for research and specialty clinical applications.The cassette dimensions are 40 x 26 x 13 mm and are available in 6 colors in quantities of 1,000.Can be used with the Mega cassettes base mold. Made of stainless steel, these reusable base molds allow easy block removal after cooling. Available in boxes of 10.
Catalog No. 50-255-2408
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.