Learn More
STEMCELL Technologies MegaCult™-C Double Chamber Slides, Size: 16 Units
SDP
Supplier:  STEMCELL Technologies 04813
Encompass_Preferred
Use MegaCult™-C Double Chamber Slides for performing colony assays in collagen gelsto detect hematopoietic progenitor cells. Available either individually or as part of the Double Chamber Slide Kit (/products/double-chamber-slide-kit.html), the slides are compatible with MegaCult™-C (/products/brands/megacult-megakaryocyte-progenitor-assay.html) and other collagen-based media. The chamber slide system allows for culture, fixation, and immunocytochemical staining of hematopoietic colony assays to be performed on the same slide. Filter Cards and Spacers (/products/filter-cards-and-spacers.html) needed for the dehydration of collagen-based gels following culture are also available for separate purchase.
Catalog No. NC1486510
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.