Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEGF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587956 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587956 Supplier Invitrogen™ Supplier No. PA587956
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, HT-29, MCF7, Mouse brain, Mouse liver, Rat brain; Cellular Location: Basolateral cell membrane, Cell membrane, Cell projection, Single-pass type I membrane protein, phagocytic cup Immunogen sequence: ADYTISGTLP HSNGGNANSH YFTNPSYHTL TQCATSPHVN NRDRMTVTKS KNNQLFVNLK NVNPGKRGPV GDCTGTLPAD WKHGGYLNEL GAFGLDRSYM GKSLKDLGKN SEYNSSNCSL SSSENPYATI KDPPV.

This gene encodes a member of the multiple epidermal growth factor-like domains protein family. The encoded protein plays a role in cell adhesion, motility and proliferation, and is a critical mediator of apoptotic cell phagocytosis as well as amyloid-beta peptide uptake in the brain. Expression of this gene may be associated with schizophrenia, and mutations in this gene are a cause of early-onset myopathy, areflexia, respiratory distress, and dysphagia (EMARDD) as well as congenital myopathy with minicores. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEGF10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MEGF10
Gene Accession No. Q6DIB5, Q96KG7
Gene Alias 3000002B06Rik; EMARDD; Gm331; KIAA1780; MEGF10; multiple EGF like domains 10; multiple EGF-like domains 10; multiple EGF-like domains protein 10; multiple EGF-like-domains 10; multiple epidermal growth factor-like domains protein 10
Gene Symbols MEGF10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 26-160 of human MEGF10 (NP_1158221).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291445, 70417, 84466
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.