Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGF11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984255
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4255 Supplier Bioss Supplier No. BS18775R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein may regulate the mosaic spacing of specific neuron subtypes in the retina through homotypic retinal neuron repulsion. Mosaics provide a mechanism to distribute each cell type evenly across the retina, ensuring that all parts of the visual field have access to a full set of processing elements.

Specifications

Antigen MEGF11
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MEGF11
Gene Accession No. A6BM72
Gene Alias 2410080H04Rik; D130061K05; Kiaa1781; Megf11; multiple EGF like domains 11; Multiple EGF-like domains protein 11; multiple EGF-like-domains 11; Multiple epidermal growth factor-like domains protein 11; UNQ1949/PRO4432
Gene Symbols MEGF11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MEGF11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84465
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.