Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEGF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562559 Supplier Invitrogen™ Supplier No. PA562559
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHEDRRGCSP LEEPMVDLDG ELPFVRPLPH IAVLQDELPQ LFQDDDVGAD EEEAELRGEH TLTEKFVCLD DSFGHDCSLT CDDC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

MEGF6 gene ontology annotations related to this gene include calcium ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEGF6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MEGF6
Gene Accession No. O75095
Gene Alias 2600001P17Rik; Egfl3; EGF-like domain-containing protein 3; EGF-like protein 3; EGF-like-domain, multiple 3; Epidermal growth factor-like protein 3; KIAA0815; MEGF6; multiple EGF like domains 6; multiple EGF-like domain protein 3; multiple EGF-like domains protein 6; multiple EGF-like-domains 6; multiple epidermal growth factor-like domains 6; multiple epidermal growth factor-like domains protein 6
Gene Symbols MEGF6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEGF6. Recombinant protein control fragment (Product #RP-100982).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.