Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Meglumine antimoniate

Catalog No. AC461140250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461140250 25 g
AC461141000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461140250 Supplier Thermo Scientific Chemicals Supplier No. 461140250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-51-7
Molecular Formula C7H17NO5·HO3Sb
Molecular Weight (g/mol) 365.97
InChI Key YPAUTKZODZLFQM-OVHUINQISA-K
PubChem CID 131664208
IUPAC Name antimony(3+);(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;trihydroxide;hydrate
SMILES CNCC(C(C(C(CO)O)O)O)O.O.[OH-].[OH-].[OH-].[Sb+3]

Specifications

Quantity 25 g
Formula Weight 365.97
Chemical Name or Material Meglumine antimoniate
Hazard Category GHS Signal Word: Warning
EINECSNumber 205-108-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.