Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meglumine Diatrizoate, Sigma-Aldrich

Catalog No. 01258882 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-882 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-882 Supplier MilliporeSigma Supelco Supplier No. M5266100G
Add to Cart
Add to Cart

Meglumine diatrizoate can be used to treat long-term Taenia saginata infection. It has also been used to tag fecal matter and bodily fluids in a study investigating preoperative examination techniques in patients with acute colon obstruction caused by colorectal cancer.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 809.13 g/mol

Specifications

Quantity 100 g
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.