Learn More
Abcam Meis2 Peptide, Synthetic, Human, Blocking, 1 mg
Description
- Peptide Type: Synthetic
- Applications: BL
- Species Reactivity: Human
- Purity: >70%
- Formulation: Liquid, pH 6.75 (0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated)
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Target Molecule | MEIS2 (Human) |
| Form | Liquid |
| Content And Storage | Store at -20°C. |
| Product Type | Blocker and Diluent |
| Buffer | 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH6.75 |
| Concentration | 1 mg/mL |
| Cross Reactivity | Human |
| Formulation | pH 6.75, 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.