Learn More
Invitrogen™ MEIS3 Monoclonal Antibody (4E12)

Description
Peptide Sequence: MARRYDELPH YPGIVDGPAA LASFPETVPA VPGPYGPHRP PQPLPPGLDS DGLKREKDEI YGHPLFPLLA LVFEKCELAT CSPRDGAGAG LGTPPGGDVC SSDSFNEDIA.
Specifications
Specifications
| Antigen | MEIS3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4E12 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | MEIS3 |
| Gene Accession No. | Q99687 |
| Gene Alias | AI573393; Homeobox protein Meis3; Meis homeobox 3; Meis homeobox 3 pseudogene 1; Meis1, myeloid ecotropic viral integration site 1 homolog 3; meis1-related protein 2; Meis3; MEIS3P1; Mrg2; myeloid ecotropic viral integration site-related gene 2 |
| Gene Symbols | MEIS3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.