Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Alm Retractors

Separates and holds tissue and organs

$564.00 - $602.00

Specifications

Product Type Alm Retractor
Grade Standard
Material Stainless Steel
Finish Matte
For Use With (Application) Separates and holds tissue and organs
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
12-460-584
View Documents
Integra™ Miltex™
MH1112
2.75 in.
Each for $564.00
Only null left
Add to Cart
 
12-460-585
View Documents
Integra™ Miltex™
MH1113
3.87 in.
Each for $602.00
Only null left
Add to Cart
 
Description

Description

  • Self-retaining
  • German stainless steel
  • Non-glare matte finish
Specifications

Specifications

Alm Retractor
Stainless Steel
Separates and holds tissue and organs
Standard
Matte
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.