Learn More
Integra™ Miltex™ MeisterHand™ Mayo Dissecting Scissors
Catalog No. 12460641
Change view
Click to view available options
Length (Metric):
13.97 cm
14.6 cm
17 cm
17.14 cm
22.86 cm
22.9 cm
Blade Style:
Curved, Beveled
Straight
Straight, Beveled
Material:
Stainless Steel
Tungsten Carbide
Product Type:
Dissecting Scissors
SuperCut Scissors
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Blade Style Material Product Type
12-460-641 17 cm Straight, Beveled Tungsten Carbide Dissecting Scissors
12-460-638 14.6 cm Curved, Beveled Stainless Steel Dissecting Scissors
12-460-639 14.6 cm Straight, Beveled Tungsten Carbide Dissecting Scissors
12-460-637 14.6 cm Straight, Beveled Stainless Steel Dissecting Scissors
12-460-640 17 cm Straight, Beveled Stainless Steel Dissecting Scissors
12-460-642 17 cm Curved, Beveled Stainless Steel Dissecting Scissors
12-460-643 22.9 cm Straight, Beveled Stainless Steel Dissecting Scissors
12-460-672 13.97 cm Straight Stainless Steel SuperCut Scissors
12-460-674 17.14 cm Straight Stainless Steel SuperCut Scissors
12-460-675 22.86 cm Straight Stainless Steel SuperCut Scissors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 12-460-641 Supplier Integra™ Miltex™ Supplier No. MH5124TC
Add to Cart
Add to Cart
distal cutting
  • Stainless steel
  • Non-glare matted finish
  • Sharp/sharp scissors available in straight and curved patterns
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue and lower blade has micro fine serrations to help prevent tissue slippage; black ring handles
Order Info
Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-sharp edge to nick or become irregular.
Specifications
Product Type Dissecting Scissors
Blade Style Straight, Beveled
Length (English) 6.75 in.
Length (Metric) 17 cm
Material Tungsten Carbide
For Use With (Application) Dissecting