Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Mayo Dissecting Scissors

Catalog No. 12460674
Change view
Click to view available options
Length (Metric):
13.97 cm
14.6 cm
17 cm
17.14 cm
22.86 cm
22.9 cm
Blade Style:
Curved, Beveled
Straight
Straight, Beveled
Material:
Stainless Steel
Tungsten Carbide
Product Type:
Dissecting Scissors
SuperCut Scissors
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Blade Style Material Product Type
12-460-674 17.14 cm Straight Stainless Steel SuperCut Scissors
12-460-638 14.6 cm Curved, Beveled Stainless Steel Dissecting Scissors
12-460-639 14.6 cm Straight, Beveled Tungsten Carbide Dissecting Scissors
12-460-637 14.6 cm Straight, Beveled Stainless Steel Dissecting Scissors
12-460-640 17 cm Straight, Beveled Stainless Steel Dissecting Scissors
12-460-642 17 cm Curved, Beveled Stainless Steel Dissecting Scissors
12-460-641 17 cm Straight, Beveled Tungsten Carbide Dissecting Scissors
12-460-643 22.9 cm Straight, Beveled Stainless Steel Dissecting Scissors
12-460-672 13.97 cm Straight Stainless Steel SuperCut Scissors
12-460-675 22.86 cm Straight Stainless Steel SuperCut Scissors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 12-460-674 Supplier Integra™ Miltex™ Supplier No. MH5SC124
Add to Cart
Add to Cart

distal cutting

  • Stainless steel
  • Non-glare matted finish
  • Sharp/sharp scissors available in straight and curved patterns
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue and lower blade has micro fine serrations to help prevent tissue slippage; black ring handles

Order Info

Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-sharp edge to nick or become irregular.

Specifications

Product Type SuperCut Scissors
Blade Style Straight
Grade Standard
Length (English) 6.75 in.
Length (Metric) 17.14 cm
Material Stainless Steel
For Use With (Application) Dissecting
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.