Learn More
Integra™ Miltex™ MeisterHand™ Metzenbaum (Lahey) Scissors
Catalog No. 12460646
Change view
Click to view available options
Blade Style:
Curved
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Style
12-460-646 Straight
12-460-647 Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-460-646 Supplier Integra™ Miltex™ Supplier No. MH5179TC
Add to Cart
Add to Cart
Standard pattern
  • Stainless steel
  • Non-glare matte finish
  • Blunt points
  • Tungsten carbide cutting edge in straight, with gold ring handles
  • Length: 5.5 in. (14cm)
Order Info
Cold or chemical sterilization not recommended for Tungsten carbide scissors.
Specifications
Blade Style Straight
Length (English) 5.625 in.
Length (Metric) 14.3 cm
For Use With (Application) Surgical, Metzenbaum
Tip Style Blunt/Blunt