Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Melan-A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583921 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583921 Supplier Invitrogen™ Supplier No. PA583921
  • $660.00
    $534.65 / Each of 1

    Save $125.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: RRRNGYRALM DKSLHVGTQC ALTRRCPQEG FDHRDSKVSL QEKNCEPVVP NAPPAYEKLS AEQSPPPYSP Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 64%.

MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melan-A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MLANA
Gene Accession No. Q16655
Gene Alias A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A
Gene Symbols MLANA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MLANA. Recombinant protein control fragment (Product #RP-98413).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.